Assay Details
Gene Symbol: ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205558.1 | NP_991121.1 | 4 - 5 | 472 | 111 | |
GenBank mRNA | BC044496.1 | - | 4 - 5 | 498 | 111 | |
BC056547.1 | - | 4 - 5 | 472 | 111 | ||
BC065471.1 | - | 4 - 5 | 472 | 111 | ||
BC155185.1 | - | 4 - 5 | 483 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
