Assay Details
Gene Symbol: gria1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205598.1 | NP_991161.1 | 10 - 11 | 1556 | 89 | |
XM_005173262.4 | XP_005173319.1 | 10 - 11 | 1654 | 89 | ||
GenBank mRNA | AF525741.1 | - | 10 - 11 | 1556 | 89 | |
BC162505.1 | - | 10 - 11 | 1492 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
