Assay Details
Gene Symbol: cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212591.1 | NP_997756.1 | 2 - 3 | 194 | 76 | |
GenBank mRNA | AF268046.1 | - | 2 - 3 | 194 | 76 | |
AY181980.1 | - | 2 - 3 | 148 | 76 | ||
BC055634.1 | - | 2 - 3 | 153 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
