Assay Details
Gene Symbol: cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212591.1 | NP_997756.1 | 3 - 4 | 273 | 106 | |
GenBank mRNA | AF268046.1 | - | 3 - 4 | 273 | 106 | |
AY181980.1 | - | 3 - 4 | 227 | 106 | ||
BC055634.1 | - | 3 - 4 | 232 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
