Assay Details
Gene Symbol: cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212591.1 | NP_997756.1 | 4 - 5 | 364 | 94 | |
GenBank mRNA | AF268046.1 | - | 4 - 5 | 364 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
