Assay Details
Gene Symbol: cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212591.1 | NP_997756.1 | 5 - 6 | 525 | 73 | |
GenBank mRNA | AF268046.1 | - | 5 - 6 | 525 | 73 | |
BC055634.1 | - | 5 - 6 | 490 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
