Assay Details
Gene Symbol: brd3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212702.1 | NP_997867.1 | 7 - 8 | 1341 | 72 | |
XM_005165593.4 | XP_005165650.2 | 7 - 8 | 1407 | 72 | ||
XM_005165594.4 | XP_005165651.2 | 7 - 8 | 1390 | 72 | ||
XM_005165595.4 | XP_005165652.2 | 7 - 8 | 1393 | 72 | ||
XM_009301936.3 | XP_009300211.1 | 7 - 8 | 1393 | 72 | ||
XM_009301938.3 | XP_009300213.1 | 7 - 8 | 1393 | 72 | ||
XM_009301940.3 | XP_009300215.1 | 7 - 8 | 1393 | 72 | ||
GenBank mRNA | BC055533.1 | - | 7 - 8 | 1407 | 72 | |
BC065949.1 | - | 7 - 8 | 1341 | 72 | ||
BC151886.1 | - | 7 - 8 | 1375 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
