Assay Details
Gene Symbol: vrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213385.1 | NP_998550.1 | 1 | 974 | 102 | |
GenBank mRNA | BC047853.1 | - | 1 | 974 | 102 | |
BC152280.1 | - | 1 | 948 | 102 |
Gene Symbol: LOC794952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021476183.1 | XP_021331858.1 | 1 | 1160 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
