Assay Details
Gene Symbol: p4ha1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214691.1 | NP_999856.1 | 7 - 8 | 1062 | 149 | |
XM_009292958.3 | XP_009291233.1 | 7 - 8 | 149 | |||
XM_009292960.3 | XP_009291235.1 | 7 - 8 | 149 | |||
XM_017351587.2 | XP_017207076.1 | 7 - 8 | 149 | |||
GenBank mRNA | BC045890.1 | - | 7 - 8 | 1062 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
