Assay Details
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631738.1 | XP_023487506.1 | 750 | 134 | ||
XM_023631739.1 | XP_023487507.1 | 469 | 134 | |||
XM_023631740.1 | XP_023487508.1 | 756 | 134 | |||
XM_023631741.1 | XP_023487509.1 | 744 | 134 | |||
XM_023631742.1 | XP_023487510.1 | 750 | 134 | |||
XM_023631743.1 | XP_023487511.1 | 232 | 134 | |||
XM_023631744.1 | XP_023487512.1 | 404 | 134 | |||
XM_023631745.1 | XP_023487513.1 | 1092 | 134 | |||
XM_023631746.1 | XP_023487514.1 | 244 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
