Assay Details
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252576.1 | NP_001239505.1 | 2 - 3 | 121 | 97 | |
GenBank mRNA | CD466901.1 | - | 2 - 3 | 121 | 97 | |
JL616180.1 | - | 2 - 3 | 115 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
