Assay Details
Gene Symbol: KHNYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623132.1 | XP_023478900.1 | 1785 | 98 | ||
XM_023623135.1 | XP_023478903.1 | 1818 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
