Assay Details
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622845.1 | XP_023478613.1 | 1776 | 74 | ||
XM_023622848.1 | XP_023478616.1 | 1761 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
