Assay Details
Gene Symbol: NUAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631468.1 | XP_023487236.1 | 836 | 65 | ||
XM_023631469.1 | XP_023487237.1 | 403 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
