Assay Details
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309219.1 | NP_001296148.1 | 1 - 2 | 213 | 82 | |
XM_005601618.3 | XP_005601675.1 | 366 | 82 | |||
XM_005601619.3 | XP_005601676.1 | 338 | 82 | |||
XM_023621981.1 | XP_023477749.1 | 487 | 82 | |||
GenBank mRNA | HQ889972.1 | - | 1 - 2 | 143 | 82 | |
JL616154.1 | - | 1 - 2 | 213 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
