Assay Details
Gene Symbol: PSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142400.1 | NP_001135872.1 | 5 - 6 | 497 | 94 | |
XM_005609945.3 | XP_005610002.1 | 521 | 94 | |||
XM_005609946.3 | XP_005610003.1 | 329 | 94 | |||
XM_023640280.1 | XP_023496048.1 | 510 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
