Assay Details
Gene Symbol: EIF4E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309215.2 | NP_001296144.1 | 4 - 5 | 430 | 87 | |
XM_005610719.3 | XP_005610776.1 | 491 | 87 | |||
XM_005610720.3 | XP_005610777.1 | 491 | 87 | |||
GenBank mRNA | CX601972.1 | - | 3 - 4 | 289 | 87 | |
DN510879.1 | - | 4 - 5 | 432 | 87 | ||
JL615897.1 | - | 4 - 5 | 416 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
