Assay Details
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309274.1 | NP_001296203.1 | 1 - 2 | 106 | 63 | |
XM_005612006.3 | XP_005612063.1 | 264 | 63 | |||
XM_023644131.1 | XP_023499899.1 | 376 | 63 | |||
GenBank mRNA | JL616152.2 | - | 1 - 2 | 106 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
