Assay Details
Gene Symbol: PTEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317260.1 | NP_001304189.1 | 6 - 7 | 1150 | 91 | |
XM_023640521.1 | XP_023496289.1 | 949 | 91 | |||
GenBank mRNA | JL615927.1 | - | 4 - 5 | 449 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
