Assay Details
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650930.1 | XP_023506698.1 | 958 | 102 | ||
GenBank mRNA | JL615946.1 | - | 6 - 7 | 578 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
