Assay Details
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917593.5 | XP_001917628.3 | 435 | 68 | ||
XM_005604669.3 | XP_005604726.1 | 347 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
