Assay Details
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163888.1 | NP_001157360.1 | 2 - 3 | 827 | 97 | |
XM_005599129.3 | XP_005599186.1 | 1174 | 97 | |||
XM_023616889.1 | XP_023472657.1 | 973 | 97 | |||
XM_023616890.1 | XP_023472658.1 | 848 | 97 | |||
GenBank mRNA | AB106114.1 | - | 1 - 2 | 138 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
