Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493570.6 | XP_001493620.1 | 4835 | 66 | ||
XM_023641680.1 | XP_023497448.1 | 4836 | 66 | |||
XM_023641681.1 | XP_023497449.1 | 4230 | 66 | |||
XM_023641682.1 | XP_023497450.1 | 4834 | 66 | |||
XM_023641683.1 | XP_023497451.1 | 4228 | 66 | |||
XM_023641684.1 | XP_023497452.1 | 4285 | 66 | |||
XM_023641685.1 | XP_023497453.1 | 1188 | 66 | |||
XM_023641687.1 | XP_023497455.1 | 459 | 66 | |||
XM_023641688.1 | XP_023497456.1 | 326 | 66 | |||
XM_023641689.1 | XP_023497457.1 | 458 | 66 | |||
GenBank mRNA | JL618455.1 | - | 2 - 3 | 708 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
