Assay Details
Gene Symbol: HM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499604.4 | XP_001499654.1 | 655 | 59 | ||
XM_005604548.3 | XP_005604605.1 | 654 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
