Assay Details
Gene Symbol: ANAPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488195.6 | XP_001488245.1 | 525 | 97 | ||
XM_023651768.1 | XP_023507536.1 | 271 | 97 | |||
XM_023651769.1 | XP_023507537.1 | 314 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
