Assay Details
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081759.1 | NP_001075228.1 | 26 - 27 | 3799 | 59 | |
XM_005597036.3 | XP_005597093.1 | 3903 | 59 | |||
GenBank mRNA | AB088366.1 | - | 26 - 27 | 3799 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
