Assay Details
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081778.2 | NP_001075247.1 | 7 - 8 | 1601 | 88 | |
GenBank mRNA | AB292109.1 | - | 7 - 8 | 1602 | 88 | |
JL616338.2 | - | 4 - 5 | 1104 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
