Assay Details
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081786.1 | NP_001075255.1 | 5 - 6 | 500 | 82 | |
GenBank mRNA | DQ385611.1 | - | 5 - 6 | 500 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
