Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081819.2 | NP_001075288.2 | 2 - 3 | 271 | 85 | |
XM_005603490.1 | XP_005603547.1 | 404 | 85 | |||
GenBank mRNA | AB035735.1 | - | 2 - 3 | 271 | 85 | |
EU438779.1 | - | 2 - 3 | 262 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
