Assay Details
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081826.3 | NP_001075295.1 | 1 - 2 | 174 | 81 | |
GenBank mRNA | AB001692.1 | - | 1 - 2 | 154 | 81 | |
CX601547.1 | - | 1 - 2 | 112 | 81 | ||
GU808339.1 | - | 1 - 2 | 175 | 81 | ||
U38956.1 | - | 1 - 2 | 75 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
