Assay Details
Gene Symbol: LITAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081861.2 | NP_001075330.2 | 2 - 3 | 379 | 86 | |
XM_023655003.1 | XP_023510771.1 | 398 | 86 | |||
GenBank mRNA | AF503366.1 | - | 3 - 4 | 412 | 86 | |
CX599393.1 | - | 1 - 2 | 148 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
