Assay Details
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081870.2 | NP_001075339.1 | 4 - 5 | 440 | 62 | |
GenBank mRNA | AJ243283.1 | - | 4 - 5 | 440 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
