Assay Details
Gene Symbol: MPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081894.2 | NP_001075363.1 | 1 - 2 | 229 | 74 | |
GenBank mRNA | AY293741.1 | - | 1 - 2 | 229 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
