Assay Details
Gene Symbol: TNNI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081904.1 | NP_001075373.1 | 6 - 7 | 497 | 71 | |
GenBank mRNA | AY819020.1 | - | 6 - 7 | 497 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
