Assay Details
Gene Symbol: TNNI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081904.1 | NP_001075373.1 | 7 - 8 | 679 | 70 | |
GenBank mRNA | AY819020.1 | - | 7 - 8 | 679 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
