Assay Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081948.1 | NP_001075417.1 | 1 - 2 | 166 | 79 | |
GenBank mRNA | AB331394.2 | - | 1 - 2 | 166 | 79 | |
AB355925.1 | - | 1 - 2 | 166 | 79 | ||
AB358958.1 | - | 1 - 2 | 166 | 79 | ||
U02929.1 | - | 1 - 2 | 166 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
