Assay Details
Gene Symbol: IFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081949.1 | NP_001075418.1 | 2 - 3 | 239 | 80 | |
GenBank mRNA | D28520.1 | - | 2 - 3 | 175 | 80 | |
EU000433.1 | - | 2 - 3 | 175 | 80 | ||
KJ814948.1 | - | 2 - 3 | 115 | 80 | ||
U04050.1 | - | 2 - 3 | 239 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
