Assay Details
Gene Symbol: IL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082500.2 | NP_001075969.2 | 6 - 7 | 672 | 95 | |
GenBank mRNA | CD466534.1 | - | 6 - 7 | 672 | 95 | |
D42146.1 | - | 5 - 6 | 624 | 95 | ||
U92480.1 | - | 6 - 7 | 651 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
