Assay Details
Gene Symbol: IL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082516.1 | NP_001075985.1 | 5 - 6 | 864 | 73 | |
GenBank mRNA | Y11129.1 | - | 864 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
