Assay Details
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082519.1 | NP_001075988.1 | 3 - 4 | 317 | 69 | |
GenBank mRNA | AF035404.1 | - | 3 - 4 | 312 | 69 | |
AF305617.1 | - | 3 - 4 | 317 | 69 | ||
EU000425.1 | - | 3 - 4 | 317 | 69 | ||
EU438769.1 | - | 3 - 4 | 317 | 69 | ||
GU139701.1 | - | 3 - 4 | 317 | 69 | ||
JQ432546.1 | - | 3 - 4 | 317 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
