Assay Details
Gene Symbol: PPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099761.1 | NP_001093231.1 | 4 - 5 | 540 | 73 | |
GenBank mRNA | EF397503.1 | - | 4 - 5 | 540 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
