Assay Details
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099767.2 | NP_001093237.1 | 3 - 4 | 355 | 93 | |
GenBank mRNA | Y18204.1 | - | 358 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
