Assay Details
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099767.2 | NP_001093237.1 | 4 - 5 | 630 | 111 | |
GenBank mRNA | JL625146.1 | - | 1 - 2 | 112 | ||
Y18204.1 | - | 633 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
