Assay Details
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114539.2 | NP_001108011.1 | 2 - 3 | 317 | 121 | |
XM_023653770.1 | XP_023509538.1 | 1466 | 121 | |||
XM_023653771.1 | XP_023509539.1 | 815 | 121 | |||
XM_023653772.1 | XP_023509540.1 | 799 | 121 | |||
XM_023653773.1 | XP_023509541.1 | 798 | 121 | |||
XM_023653774.1 | XP_023509542.1 | 503 | 121 | |||
XM_023653775.1 | XP_023509543.1 | 488 | 121 | |||
XM_023653776.1 | XP_023509544.1 | 480 | 121 | |||
XM_023653777.1 | XP_023509545.1 | 400 | 121 | |||
XM_023653778.1 | XP_023509546.1 | 345 | 121 | |||
XM_023653779.1 | XP_023509547.1 | 695 | 121 | |||
XM_023653780.1 | XP_023509548.1 | 455 | 121 | |||
XM_023653781.1 | XP_023509549.1 | 386 | 121 | |||
XM_023653782.1 | XP_023509550.1 | 404 | 121 | |||
XM_023653783.1 | XP_023509551.1 | 335 | 121 | |||
GenBank mRNA | AB915864.1 | - | 3 - 4 | 376 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
