Assay Details
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115158.1 | NP_001108630.1 | 4 - 5 | 494 | 144 | |
GenBank mRNA | AB040095.1 | - | 2 - 3 | 151 | 144 | |
CX603716.1 | - | 4 - 5 | 529 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
