Assay Details
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163859.1 | NP_001157331.1 | 1 - 2 | 1167 | 79 | |
GenBank mRNA | AF167158.1 | - | 1 - 2 | 760 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
