Assay Details
Gene Symbol: PLAUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163881.2 | NP_001157353.2 | 6 - 7 | 900 | 56 | |
GenBank mRNA | AF230359.1 | - | 1 - 2 | 25 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
