Assay Details
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163883.1 | NP_001157355.1 | 2 | 630 | 91 | |
GenBank mRNA | AJ439891.1 | - | 1 | 153 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
