Assay Details
Gene Symbol: NMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242511.1 | NP_001229440.1 | 2 - 3 | 182 | 77 | |
XM_005601393.3 | XP_005601450.1 | 465 | 77 | |||
GenBank mRNA | AY246724.1 | - | 2 - 3 | 122 | 77 | |
JL618615.1 | - | 2 - 3 | 286 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
