Assay Details
Gene Symbol: HTR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301236.1 | NP_001288165.1 | 4 - 5 | 420 | 64 | |
GenBank mRNA | AY263356.1 | - | 1 - 2 | 109 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
